siRNA Details
Gene Symbol: A430106F12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020773.1 | - | 1 | 532 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724657.1 | - | 1 | 533 | Chr. 13 - 117265024 - 117265597 [+] on Build GRCm38 |
