siRNA Details
Gene Symbol: Gm19863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157548.1 | - | 1 | 2182 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY571336.1 | - | 1 | 514 | Chr. 1 - 37694447 - 37694992 [-] on Build GRCm38 |
