siRNA Details
Gene Symbol: 1110001D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK003212.1 | - | 1 | 304 | ||
AV006698.1 | - | 1 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY703524.1 | - | 1 | 302 | Chr. 4 - 151932378 - 151932849 [-] on Build GRCm38 |
