siRNA Details
Gene Symbol: 4930556M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045063.1 | 7 | 2405 | |||
GenBank mRNA | AK030258.1 | - | 7 | 2406 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930556M19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045063.1 | 7 | 2405 | |||
GenBank mRNA | AK030258.1 | - | 7 | 2406 |