siRNA Details
Gene Symbol: 4930404I20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015082.1 | - | 1 | 27 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714612.1 | - | 1 | 27 | Chr. 6 - 139555559 - 139557611 [-] on Build GRCm38 |
