siRNA Details
Gene Symbol: 4930572P05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016285.1 | - | 1 | 40 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715908.1 | - | 1 | 37 | Chr. 16 - 78454713 - 78455335 [+] on Build GRCm38 |
