siRNA Details
Gene Symbol: D730003I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021325.1 | - | 2 | 502 | ||
AK157967.1 | - | 1 | 1167 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY766140.1 | - | 1 | 290 | Chr. 1 - 191225349 - 191226045 [+] on Build GRCm38 |
