siRNA Details
Gene Symbol: 4930572K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045371.1 | 1, 2 | 54 | |||
GenBank mRNA | AK016281.1 | - | 1, 2 | 55 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715905.1 | - | 1, 2 | 55 | Chr. 5 - 127170565 - 127184335 [+] on Build GRCm38 |
