siRNA Details
Gene Symbol: 4930551E15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019758.1 | - | 1 | 312 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715694.1 | - | 1 | 312 | Chr. 7 - 124186727 - 124187128 [+] on Build GRCm38 |
