siRNA Details
Gene Symbol: 4930551I20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019759.1 | - | 1 | 166 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715697.1 | - | 1 | 166 | Chr. 14 - 60399137 - 60399465 [+] on Build GRCm38 |
