siRNA Details
Gene Symbol: 2900056L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013704.1 | - | 1 | 190 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY712683.1 | - | 1 | 186 | Chr. 15 - 62259296 - 62260215 [+] on Build GRCm38 |
