siRNA Details
Gene Symbol: 4930524B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015880.1 | - | 3 | 201 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715434.1 | - | 3 | 201 | Chr. 6 - 19249065 - 19278843 [-] on Build GRCm38 |
