siRNA Details
Gene Symbol: 4930547M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016061.1 | - | 2 | 376 | ||
AK036704.1 | - | 2 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715658.1 | - | 2 | 372 | Chr. 10 - 39899348 - 39906111 [+] on Build GRCm38 |
