siRNA Details
Gene Symbol: 4930548F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016066.1 | - | 1 | 120 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715663.1 | - | 1 | 120 | Chr. 13 - 31811162 - 31811988 [+] on Build GRCm38 |
