siRNA Details
Gene Symbol: Gm11186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046047.1 | 4 | 809 | |||
GenBank mRNA | AK043416.1 | - | 4 | 809 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB260379.2 | - | 1 | 403 | Chr. 11 - 53111454 - 53112197 [-] on Build GRCm38 |
