siRNA Details
Gene Symbol: Gm20754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040557.1 | 3 | 411 | |||
GenBank mRNA | AK044425.1 | - | 3 | 412 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB642374.1 | - | 3 | 412 | Chr. 3 - 73066351 - 73565177 [+] on Build GRCm38 |
