siRNA Details
Gene Symbol: Gm20754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040557.1 | 4 | 706 | |||
GenBank mRNA | AK044425.1 | - | 4 | 707 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040557.1 | 4 | 706 | |||
GenBank mRNA | AK044425.1 | - | 4 | 707 |