siRNA Details
Gene Symbol: Gm20754 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040557.1 | 5 | 1020 | |||
GenBank mRNA | AK044425.1 | - | 5 | 1021 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB278646.2 | - | 2 | 342 | Chr. 3 - 73565192 - 73594830 [+] on Build GRCm38 |
