siRNA Details
Gene Symbol: 1810012K16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045473.1 | 2 | 371 | |||
GenBank mRNA | AK007460.1 | - | 2 | 372 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707529.1 | - | 2 | 372 | Chr. 8 - 22398721 - 22399594 [+] on Build GRCm38 |
