siRNA Details
Gene Symbol: Gm16152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048050.1 | - | 156 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB648653.1 | - | 1 | 153 | Chr. 1 - 38384951 - 38391085 [+] on Build GRCm38 |
