siRNA Details
Gene Symbol: Gm16152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048050.1 | - | 854 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB366045.2 | - | 2 | 366 | Chr. 1 - 38390890 - 38393974 [+] on Build GRCm38 |
