siRNA Details
Gene Symbol: Gm13483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040362.1 | 2 | 122 | |||
GenBank mRNA | AK045825.1 | - | 2 | 68 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB644362.1 | - | 2 | 68 | Chr. 2 - 50296865 - 50433981 [+] on Build GRCm38 |
