siRNA Details
Gene Symbol: Gm13848 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085570.1 | - | 2 | 395 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661489.1 | - | 2 | 388 | Chr. 6 - 31663620 - 31666111 [-] on Build GRCm38 |
