siRNA Details
Gene Symbol: Myl6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017625.1 | - | 1 | 656 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DV047970.1 | - | 1 | 224 | Chr. 10 - 128494763 - 128495555 [+] on Build GRCm38 |
