siRNA Details
Gene Symbol: 5730435O14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045341.1 | 3, 4 | 500 | |||
GenBank mRNA | AK017620.1 | - | 3, 4 | 500 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717398.1 | - | 3, 4 | 500 | Chr. 10 - 41568016 - 41576482 [+] on Build GRCm38 |
