siRNA Details
Gene Symbol: 1700034G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045396.1 | 5 | 630 | |||
NR_045397.1 | 5 | 599 | ||||
GenBank mRNA | AK006334.1 | - | 5 | 600 | ||
AK006593.1 | - | 5 | 631 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706406.1 | - | 5 | 600 | Chr. 5 - 112583317 - 112589996 [-] on Build GRCm38 |
