siRNA Details
Gene Symbol: 1700034G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045396.1 | 2 | 201 | |||
NR_045397.1 | 2 | 201 | ||||
GenBank mRNA | AK006334.1 | - | 2 | 202 | ||
AK006593.1 | - | 2 | 202 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700034G24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045396.1 | 2 | 201 | |||
NR_045397.1 | 2 | 201 | ||||
GenBank mRNA | AK006334.1 | - | 2 | 202 | ||
AK006593.1 | - | 2 | 202 |