siRNA Details
Gene Symbol: 1700025F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040578.1 | 4 | 587 | |||
GenBank mRNA | AK006336.1 | - | 4 | 591 | ||
AK015773.1 | - | 4 | 566 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715316.1 | - | 4 | 548 | Chr. 10 - 119413483 - 119426839 [+] on Build GRCm38 |
