siRNA Details
Gene Symbol: 1700025F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040578.1 | 4 | 821 | |||
GenBank mRNA | AK006336.1 | - | 4 | 825 | ||
AK015773.1 | - | 4 | 800 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700025F24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040578.1 | 4 | 821 | |||
GenBank mRNA | AK006336.1 | - | 4 | 825 | ||
AK015773.1 | - | 4 | 800 |