siRNA Details
Gene Symbol: 1700029B24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006483.1 | - | 1 | 240 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706522.1 | - | 1 | 240 | Chr. 3 - 10241812 - 10242308 [+] on Build GRCm38 |
