siRNA Details
Gene Symbol: Gm32014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110513.1 | 5 | 3719 | |||
GenBank mRNA | AK054106.1 | - | 5 | 3719 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB549988.2 | - | 1 | 220 | Chr. 9 - 8644073 - 8644741 [-] on Build GRCm38 |
