siRNA Details
Gene Symbol: 2010203P06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008426.1 | - | 2 | 226 | ||
AK008432.1 | - | 2 | 226 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708335.1 | - | 2 | 226 | Chr. 13 - 56595537 - 56596642 [+] on Build GRCm38 |
