siRNA Details
Gene Symbol: 1700006P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005685.1 | - | 1 | 84 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705863.1 | - | 1 | 80 | Chr. 1 - 137325433 - 137325839 [-] on Build GRCm38 |
