siRNA Details
Gene Symbol: Fendrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004221.1 | - | 3 | 880 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB365376.2 | - | 2 | 483 | Chr. 8 - 121058803 - 121059584 [-] on Build GRCm38 |
