siRNA Details
Gene Symbol: 4930448I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040475.1 | 1 | 60 | |||
GenBank mRNA | AK015419.1 | - | 1 | 61 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714967.1 | - | 1 | 61 | Chr. 1 - 41167560 - 41181252 [-] on Build GRCm38 |
