siRNA Details
Gene Symbol: Gm36529 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK049017.1 | - | 4 | 564 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB650341.1 | - | 4 | 564 | Chr. 13 - 70840451 - 70858713 [+] on Build GRCm38 |
