siRNA Details
Gene Symbol: Pantr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048630.1 | - | 2 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB649785.1 | - | 2 | 357 | Chr. 1 - 42693976 - 42694825 [-] on Build GRCm38 |
