siRNA Details
Gene Symbol: A830008E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137140.1 | - | 1 | 349 | ||
AK154552.1 | - | 4 | 3156 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: A830008E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK137140.1 | - | 1 | 349 | ||
AK154552.1 | - | 4 | 3156 |