siRNA Details
Gene Symbol: A230001M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040391.1 | 2, 3 | 640 | |||
GenBank mRNA | AK038444.1 | - | 2, 3 | 524 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632192.1 | - | 2, 3 | 524 | Chr. 3 - 102262522 - 102278973 [+] on Build GRCm38 |
