siRNA Details
Gene Symbol: Gm29685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK147773.1 | - | 138 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY740883.1 | - | 1, 2 | 135 | Chr. 10 - 107946647 - 107976215 [+] on Build GRCm38 |
