siRNA Details
Gene Symbol: 4833404L02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014654.1 | - | 1 | 853 | ||
AK029357.1 | - | 1 | 852 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV248175.2 | - | 1 | 936 | Chr. 7 - 17058566 - 17059423 [+] on Build GRCm38 |
