siRNA Details
Gene Symbol: Gm34433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC044776.2 | - | 2 | 109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI732276.1 | - | 2 | 110 | Chr. 7 - 19333129 - 19339181 [+] on Build GRCm38 |
