siRNA Details
Gene Symbol: 4921531P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045361.1 | 3 | 339 | |||
GenBank mRNA | AK007141.1 | - | 3 | 340 | ||
AK014992.1 | - | 4 | 1151 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707224.1 | - | 3 | 340 | Chr. 18 - 83292792 - 83325884 [-] on Build GRCm38 |
