siRNA Details
Gene Symbol: Gm36799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039939.1 | - | 57 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB634609.1 | - | 1 | 54 | Chr. 9 - 43376627 - 43377483 [+] on Build GRCm38 |
