siRNA Details
Gene Symbol: 2310065F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079404.1 | - | 1 | 52 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB629401.1 | - | 1 | 49 | Chr. 11 - 67116112 - 67116839 [-] on Build GRCm38 |
