siRNA Details
Gene Symbol: Gm3510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK138389.1 | - | 456 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY724079.1 | - | 2 | 452 | Chr. 5 - 15924555 - 15933403 [-] on Build GRCm38 |
