siRNA Details
Gene Symbol: Gm40761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142676.1 | - | 2 | 423 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730932.1 | - | 2 | 418 | Chr. 10 - 111337128 - 111346722 [+] on Build GRCm38 |
