siRNA Details
Gene Symbol: 8030448I15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020202.1 | - | 1 | 237 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720425.1 | - | 1 | 234 | Chr. 1 - 157156086 - 157156666 [-] on Build GRCm38 |
