siRNA Details
Gene Symbol: 8030450C14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020204.1 | - | 1 | 195 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720428.1 | - | 1 | 191 | Chr. 10 - 61931660 - 61932320 [-] on Build GRCm38 |
