siRNA Details
Gene Symbol: 8030455M16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020209.1 | - | 2 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY720434.1 | - | 2 | 337 | Chr. 8 - 110047432 - 110056940 [+] on Build GRCm38 |
